Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291225.1 | NP_001278154.1 | 5 - 6 | 1452 | 76 | |
NM_021286.4 | NP_067261.2 | 5 - 6 | 1452 | 76 | ||
GenBank mRNA | AB206791.1 | - | 5 - 6 | 1388 | 76 | |
AK147705.1 | - | 5 - 6 | 1371 | 76 | ||
AK165757.1 | - | 5 - 6 | 1236 | 76 | ||
AK220513.1 | - | 4 - 5 | 2082 | 76 | ||
BC053011.1 | - | 5 - 6 | 1348 | 5698186 | 76 | |
BC055345.1 | - | 5 - 6 | 1452 | 6833344 | 76 | |
D29763.1 | - | 5 - 6 | 1279 | 76 | ||
D64009.1 | - | 5 - 6 | 1252 | 76 | ||
D64010.1 | - | 5 - 6 | 1252 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |