Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023047.2 | NP_075534.1 | 3 - 4 | 701 | 76 | |
GenBank mRNA | AF249295.1 | - | 3 - 4 | 865 | 76 | |
AK044866.1 | - | 3 - 4 | 573 | 76 | ||
BC065054.1 | - | 3 - 4 | 701 | 5709296 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |