Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF040094.2 | - | 9 - 10 | 1168 | 109 | |
AK004601.1 | - | 10 - 11 | 1175 | 109 | ||
AK004722.2 | - | 9 - 10 | 1070 | 109 | ||
AY007563.1 | - | 15 - 16 | 1900 | 109 | ||
BC028864.1 | - | 15 - 16 | 1886 | 4457437 | 109 | |
BE996479.1 | - | 3 - 4 | 323 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |