Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 8 - 9 | 1481 | 75 | |
GenBank mRNA | AK084837.1 | - | 8 - 9 | 1458 | 75 | |
AK135179.1 | - | 8 - 9 | 1429 | 75 | ||
AK135430.1 | - | 8 - 9 | 975 | 75 | ||
AK171938.1 | - | 8 - 9 | 1392 | 75 | ||
BC050946.1 | - | 8 - 9 | 1419 | 6822283 | 75 | |
BC054537.1 | - | 8 - 9 | 1403 | 75 | ||
D12646.1 | - | 8 - 9 | 1808 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |