Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277928.1 | NP_001264857.1 | 6 - 7 | 699 | 84 | |
NM_008484.2 | NP_032510.2 | 6 - 7 | 738 | 84 | ||
GenBank mRNA | AK155769.1 | - | 1551 | 84 | ||
BC008516.1 | - | 6 - 7 | 699 | 3592888 | 84 | |
BY248755.1 | - | 3 - 4 | 288 | 84 | ||
U43298.1 | - | 6 - 7 | 714 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |