Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281980.1 | NP_001268909.1 | 16 - 17 | 1344 | 83 | |
NM_001281981.1 | NP_001268910.1 | 17 - 18 | 1385 | 83 | ||
NM_008511.3 | NP_032537.3 | 16 - 17 | 1616 | 83 | ||
GenBank mRNA | AK153865.1 | - | 16 - 17 | 1346 | 83 | |
AK160009.1 | - | 16 - 17 | 1576 | 83 | ||
AK166635.1 | - | 14 - 15 | 1065 | 83 | ||
AK168775.1 | - | 14 - 15 | 1042 | 83 | ||
AK169578.1 | - | 17 - 18 | 1369 | 83 | ||
AY753324.1 | - | 15 - 16 | 1179 | 83 | ||
BC052909.1 | - | 16 - 17 | 1552 | 30076553 | 83 | |
U10484.1 | - | 16 - 17 | 1322 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |