Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011716.2 | NP_035846.1 | 6 - 7 | 840 | 88 | |
GenBank mRNA | AF084482.1 | - | 6 - 7 | 773 | 88 | |
AJ011971.1 | - | 6 - 7 | 767 | 88 | ||
AK144561.1 | - | 6 - 7 | 792 | 88 | ||
AK145023.1 | - | 6 - 7 | 812 | 88 | ||
AK170180.1 | - | 6 - 7 | 795 | 88 | ||
AK185636.1 | - | 2 - 3 | 263 | 88 | ||
BC046988.1 | - | 6 - 7 | 776 | 6492038 | 88 | |
CX562372.1 | - | 4 - 5 | 470 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |