Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146087.1 | NP_001139559.1 | 3 - 4 | 345 | 89 | |
NM_001146088.1 | NP_001139560.1 | 2 - 3 | 219 | 89 | ||
NM_019451.2 | NP_062324.2 | 1 - 2 | 256 | 89 | ||
GenBank mRNA | AJ250429.1 | - | 1 - 2 | 137 | 89 | |
AK008977.1 | - | 2 - 3 | 157 | 89 | ||
AK009741.1 | - | 1 - 2 | 258 | 89 | ||
AK010020.1 | - | 1 - 2 | 255 | 89 | ||
AK014576.2 | - | 345 | 89 | |||
BC132600.1 | - | 1 - 2 | 133 | 40130877 | 89 | |
BC132602.1 | - | 1 - 2 | 133 | 40130879 | 89 | |
BY109350.1 | - | 2 - 3 | 160 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |