Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020277.2 | NP_064673.2 | 12 - 13 | 1829 | 59 | |
GenBank mRNA | AB039952.1 | - | 11 - 12 | 1746 | 59 | |
AF228681.1 | - | 12 - 13 | 1829 | 59 | ||
AJ271092.2 | - | 11 - 12 | 1742 | 59 | ||
AK035197.1 | - | 8 - 9 | 1359 | 59 | ||
AK044908.1 | - | 8 - 9 | 1444 | 59 | ||
AK161030.1 | - | 3 - 4 | 422 | 59 | ||
AY280364.1 | - | 12 - 13 | 1793 | 59 | ||
AY280365.1 | - | 12 - 13 | 1793 | 59 | ||
BC133712.1 | - | 11 - 12 | 1746 | 40143371 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |