Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB162929.1 | - | 3 - 4 | 422 | 53 | |
AF190699.1 | - | 3 - 4 | 494 | 53 | ||
AF190991.1 | - | 3 - 4 | 494 | 53 | ||
AK018148.1 | - | 3 - 4 | 486 | 53 | ||
AK031550.1 | - | 3 - 4 | 522 | 53 | ||
AK042758.1 | - | 3 - 4 | 275 | 53 | ||
AK043998.1 | - | 3 - 4 | 542 | 53 | ||
AK048413.1 | - | 3 - 4 | 493 | 53 | ||
AK049903.1 | - | 2 - 3 | 228 | 53 | ||
AK081981.1 | - | 3 - 4 | 495 | 53 | ||
AK083126.1 | - | 2 - 3 | 619 | 53 | ||
BC046310.1 | - | 3 - 4 | 480 | 6307420 | 53 | |
BC065694.1 | - | 3 - 4 | 486 | 6813365 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |