Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024188.6 | NP_077150.1 | 12 - 13 | 1512 | 73 | |
GenBank mRNA | AB085609.1 | - | 12 - 13 | 1254 | 73 | |
AK009027.1 | - | 12 - 13 | 1292 | 73 | ||
AK011354.1 | - | 12 - 13 | 1192 | 73 | ||
AK078086.1 | - | 12 - 13 | 1510 | 73 | ||
AK146158.1 | - | 12 - 13 | 73 | |||
AK146342.1 | - | 12 - 13 | 1257 | 73 | ||
AK151662.1 | - | 12 - 13 | 1251 | 73 | ||
AK151695.1 | - | 12 - 13 | 1242 | 73 | ||
AK167681.1 | - | 12 - 13 | 1342 | 73 | ||
AK217531.1 | - | 2 - 3 | 87 | 73 | ||
BC003422.1 | - | 12 - 13 | 1216 | 2598824 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |