Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162946.1 | NP_001156418.1 | 9 - 10 | 1170 | 56 | |
NM_008797.3 | NP_032823.2 | 8 - 9 | 1147 | 56 | ||
GenBank mRNA | AK128927.1 | - | 1097 | 56 | ||
AK148328.1 | - | 9 - 10 | 1117 | 56 | ||
AK170596.1 | - | 9 - 10 | 1130 | 56 | ||
AK170871.1 | - | 9 - 10 | 1120 | 56 | ||
AK172320.1 | - | 9 - 10 | 1098 | 56 | ||
BC055030.1 | - | 9 - 10 | 1105 | 5053475 | 56 | |
L09192.1 | - | 8 - 9 | 1123 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |