Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019764.2 | NP_062738.2 | 6 - 7 | 1721 | 128 | |
GenBank mRNA | AF175967.1 | - | 6 - 7 | 1004 | 128 | |
AF175968.1 | - | 4 - 5 | 1861 | 128 | ||
AK040912.1 | - | 6 - 7 | 1802 | 128 | ||
AK077535.1 | - | 6 - 7 | 1717 | 128 | ||
AK141348.1 | - | 6 - 7 | 1721 | 128 | ||
AK164279.1 | - | 6 - 7 | 1790 | 128 | ||
AK164819.1 | - | 6 - 7 | 1721 | 128 | ||
AK169419.1 | - | 6 - 7 | 1721 | 128 | ||
AX763686.1 | - | 5 - 6 | 1560 | 128 | ||
BC027824.1 | - | 6 - 7 | 1691 | 5323888 | 128 | |
BC035201.1 | - | 6 - 7 | 1703 | 128 | ||
BC108411.1 | - | 6 - 7 | 1755 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |