Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164600.1 | NP_001158072.1 | 3 - 4 | 271 | 94 | |
NM_028364.2 | NP_082640.2 | 3 - 4 | 301 | 94 | ||
NM_133691.5 | NP_598452.2 | 3 - 4 | 301 | 94 | ||
GenBank mRNA | AK012434.1 | - | 3 - 4 | 241 | 94 | |
AK075960.1 | - | 3 - 4 | 269 | 94 | ||
AK146534.1 | - | 3 - 4 | 271 | 94 | ||
AK149632.1 | - | 3 - 4 | 267 | 94 | ||
AK151526.1 | - | 3 - 4 | 265 | 94 | ||
AK168789.1 | - | 3 - 4 | 268 | 94 | ||
AK180376.1 | - | 2 - 3 | 143 | 94 | ||
AK185614.1 | - | 2 - 3 | 143 | 94 | ||
AK189651.1 | - | 2 - 3 | 143 | 94 | ||
AK209860.1 | - | 2 - 3 | 146 | 94 | ||
BC010601.1 | - | 3 - 4 | 271 | 3257153 | 94 | |
BY296547.1 | - | 3 - 4 | 302 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |