Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080968.1 | NP_001074437.1 | 7 - 8 | 601 | 109 | |
NM_133852.2 | NP_598613.2 | 8 - 9 | 682 | 109 | ||
GenBank mRNA | AK076370.1 | - | 3 - 4 | 222 | 109 | |
AK077516.1 | - | 7 - 8 | 602 | 109 | ||
AK220509.1 | - | 8 - 9 | 610 | 109 | ||
BC011407.1 | - | 6 - 7 | 402 | 3596079 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |