Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164433.1 | NP_001157905.1 | 1 - 2 | 225 | 63 | |
NM_138315.2 | NP_612188.1 | 1 - 2 | 225 | 63 | ||
GenBank mRNA | AK154665.1 | - | 1 - 2 | 165 | 63 | |
AK154930.1 | - | 1 - 2 | 205 | 63 | ||
AK157196.1 | - | 1 - 2 | 164 | 63 | ||
AK159369.1 | - | 1 - 2 | 172 | 63 | ||
AK170382.1 | - | 1 - 2 | 164 | 63 | ||
AK170827.1 | - | 1 - 2 | 196 | 63 | ||
AK170874.1 | - | 1 - 2 | 207 | 63 | ||
AK170881.1 | - | 1 - 2 | 191 | 63 | ||
AK171234.1 | - | 1 - 2 | 192 | 63 | ||
AK171411.1 | - | 1 - 2 | 226 | 63 | ||
AK171429.1 | - | 1 - 2 | 204 | 63 | ||
AK171479.1 | - | 1 - 2 | 198 | 63 | ||
AK171779.1 | - | 1 - 2 | 204 | 63 | ||
AK172562.1 | - | 1 - 2 | 198 | 63 | ||
AK172716.1 | - | 1 - 2 | 151 | 63 | ||
BC021477.1 | - | 1 - 2 | 173 | 5343552 | 63 | |
BC034682.1 | - | 1 - 2 | 117 | 3485807 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |