Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138756.4 | NP_620095.1 | 5 - 6 | 549 | 100 | |
GenBank mRNA | AK042075.1 | - | 5 - 6 | 638 | 100 | |
AK053646.1 | - | 5 - 6 | 665 | 100 | ||
AK078779.1 | - | 5 - 6 | 550 | 100 | ||
BC008171.1 | - | 5 - 6 | 541 | 3710468 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |