Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145220.2 | NP_660255.1 | 17 - 18 | 1711 | 76 | |
GenBank mRNA | AK160075.1 | - | 17 - 18 | 1690 | 76 | |
AK160105.1 | - | 17 - 18 | 1687 | 76 | ||
AK168403.1 | - | 17 - 18 | 1674 | 76 | ||
AK168748.1 | - | 17 - 18 | 1667 | 76 | ||
AK171512.1 | - | 17 - 18 | 1667 | 76 | ||
AY113706.1 | - | 17 - 18 | 1713 | 76 | ||
BC002232.1 | - | 5 - 6 | 522 | 76 | ||
BC048906.1 | - | 17 - 18 | 1649 | 6475576 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |