Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027188.3 | NP_081464.1 | 10 - 11 | 1128 | 57 | |
GenBank mRNA | AK010447.1 | - | 10 - 11 | 1162 | 57 | |
AK043988.1 | - | 10 - 11 | 1138 | 57 | ||
AK044168.1 | - | 6 - 7 | 689 | 57 | ||
AK046829.1 | - | 7 - 8 | 735 | 57 | ||
AK163057.1 | - | 10 - 11 | 1145 | 57 | ||
AK167875.1 | - | 10 - 11 | 1143 | 57 | ||
BC019663.1 | - | 10 - 11 | 57 | |||
BC038482.1 | - | 10 - 11 | 57 | |||
BC052431.1 | - | 10 - 11 | 1128 | 6837012 | 57 | |
BC061485.1 | - | 10 - 11 | 1111 | 5370924 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |