Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277296.1 | NP_001264225.1 | 2 - 3 | 413 | 111 | |
NM_001277297.1 | NP_001264226.1 | 2 - 3 | 225 | 111 | ||
NM_028105.4 | NP_082381.1 | 2 - 3 | 334 | 111 | ||
GenBank mRNA | AK011310.1 | - | 2 - 3 | 185 | 111 | |
AK050868.1 | - | 2 - 3 | 315 | 111 | ||
AK154839.1 | - | 2 - 3 | 280 | 111 | ||
BC010539.1 | - | 2 - 3 | 227 | 4162365 | 111 | |
BY291967.1 | - | 2 - 3 | 335 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |