Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sohlh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028937.3 | NP_083213.2 | 6 - 7 | 657 | 73 | |
GenBank mRNA | AK016704.1 | - | 6 - 7 | 659 | 73 | |
BC120917.1 | - | 6 - 7 | 652 | 40092728 | 73 | |
BC120918.2 | - | 6 - 7 | 652 | 40092729 | 73 | |
DQ086115.1 | - | 6 - 7 | 657 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |