Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029492.4 | NP_083768.4 | 1 - 2 | 200 | 72 | |
GenBank mRNA | AK017158.1 | - | 1 - 2 | 201 | 72 | |
AK031184.1 | - | 1 - 2 | 159 | 72 | ||
AK049270.1 | - | 1 - 2 | 180 | 72 | ||
AK139216.1 | - | 1 - 2 | 159 | 72 | ||
AK140182.1 | - | 1 - 2 | 161 | 72 | ||
AK151840.1 | - | 1 - 2 | 215 | 72 | ||
AK163174.1 | - | 1 - 2 | 198 | 72 | ||
AK170017.1 | - | 1 - 2 | 218 | 72 | ||
AK170139.1 | - | 1 - 2 | 141 | 72 | ||
AY668951.1 | - | 1 - 2 | 118 | 72 | ||
BC019536.1 | - | 1 - 2 | 217 | 4039734 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |