Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165991.1 | NP_001159463.1 | 4 - 5 | 474 | 101 | |
NM_001165992.1 | NP_001159464.1 | 4 - 5 | 455 | 101 | ||
NM_001165995.1 | NP_001159467.1 | 3 - 4 | 361 | 101 | ||
NM_029702.4 | NP_083978.3 | 4 - 5 | 468 | 101 | ||
NR_028585.1 | 4 - 5 | 468 | 101 | |||
GenBank mRNA | AJ413953.1 | - | 4 - 5 | 421 | 101 | |
AK005174.1 | - | 4 - 5 | 416 | 101 | ||
AK044759.1 | - | 4 - 5 | 426 | 101 | ||
AK050136.1 | - | 1 - 2 | 1326 | 101 | ||
AK185500.1 | - | 1 - 2 | 74 | 101 | ||
BC010713.1 | - | 4 - 5 | 415 | 2649462 | 101 | |
BC046782.1 | - | 4 - 5 | 406 | 5720540 | 101 | |
BC046786.1 | - | 4 - 5 | 406 | 6416321 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |