Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadsyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ305343.1 | - | 18 - 19 | 1867 | 61 | |
AK020264.1 | - | 18 - 19 | 1867 | 61 | ||
AK041101.1 | - | 18 - 19 | 1854 | 61 | ||
AK042389.1 | - | 18 - 19 | 1827 | 61 | ||
BC018248.1 | - | 16 - 17 | 1633 | 61 | ||
BC038016.1 | - | 18 - 19 | 1813 | 3589586 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |