Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008027.2 | NP_032053.1 | 4 - 5 | 371 | 107 | |
GenBank mRNA | AF145044.1 | - | 3 - 4 | 212 | 107 | |
AK150692.1 | - | 4 - 5 | 393 | 107 | ||
AK150827.1 | - | 4 - 5 | 320 | 107 | ||
AK167323.1 | - | 4 - 5 | 298 | 107 | ||
AY167925.1 | - | 4 - 5 | 377 | 107 | ||
BC004647.1 | - | 4 - 5 | 381 | 3158071 | 107 | |
U90435.1 | - | 4 - 5 | 371 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |