Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204973.1 | NP_001191902.1 | 10 - 11 | 3557 | 97 | |
NM_010238.3 | NP_034368.2 | 10 - 11 | 3376 | 97 | ||
NR_037970.1 | 10 - 11 | 3203 | 97 | |||
GenBank mRNA | AB212273.1 | - | 8 - 9 | 1711 | 97 | |
AF045462.1 | - | 9 - 10 | 2522 | 97 | ||
AK158970.1 | - | 10 - 11 | 3377 | 97 | ||
AK168525.1 | - | 10 - 11 | 3248 | 97 | ||
AK220444.1 | - | 9 - 10 | 2524 | 97 | ||
BC046809.1 | - | 10 - 11 | 2238 | 97 | ||
BC138655.1 | - | 9 - 10 | 2150 | 8861675 | 97 | |
BC138656.1 | - | 9 - 10 | 2150 | 8861676 | 97 | |
KJ650222.1 | - | 9 - 10 | 1842 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |