Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017383.3 | NP_059079.2 | 18 - 19 | 2716 | 115 | |
GenBank mRNA | AB032602.1 | - | 17 - 18 | 2402 | 115 | |
AK052972.1 | - | 19 - 20 | 2609 | 115 | ||
BC076594.1 | - | 18 - 19 | 2716 | 30649661 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |