Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276358.1 | NP_001263287.1 | 4 - 5 | 357 | 65 | |
NM_030716.3 | NP_109641.2 | 5 - 6 | 692 | 65 | ||
NM_145703.2 | NP_663749.1 | 6 - 7 | 746 | 65 | ||
NM_145704.2 | NP_663750.1 | 4 - 5 | 596 | 65 | ||
GenBank mRNA | AF439339.1 | - | 6 - 7 | 486 | 65 | |
AF439340.1 | - | 5 - 6 | 432 | 65 | ||
AF439341.1 | - | 4 - 5 | 336 | 65 | ||
AJ278535.1 | - | 5 - 6 | 282 | 65 | ||
AJ278536.1 | - | 6 - 7 | 432 | 65 | ||
AJ278537.1 | - | 5 - 6 | 378 | 65 | ||
AY647241.1 | - | 4 - 5 | 357 | 65 | ||
BC056434.1 | - | 5 - 6 | 1463 | 65 | ||
BC132215.1 | - | 3 - 4 | 370 | 40130492 | 65 | |
BC138496.1 | - | 4 - 5 | 412 | 8861516 | 65 | |
DQ148496.1 | - | 6 - 7 | 486 | 65 | ||
DQ148497.1 | - | 5 - 6 | 432 | 65 | ||
DQ148498.1 | - | 4 - 5 | 336 | 65 | ||
DQ148505.1 | - | 4 - 5 | 354 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |