Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145384.2 | NP_663359.2 | 6 - 7 | 797 | 111 | |
GenBank mRNA | AK081661.1 | - | 6 - 7 | 784 | 111 | |
AK156668.1 | - | 6 - 7 | 797 | 111 | ||
BC019216.1 | - | 6 - 7 | 768 | 5065163 | 111 | |
BC025609.1 | - | 5 - 6 | 735 | 5309871 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |