Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130194.1 | NP_001123666.1 | 3 - 4 | 336 | 56 | |
NR_052011.1 | 3 - 4 | 577 | 56 | |||
GenBank mRNA | AY450428.1 | - | 3 - 4 | 578 | 56 | |
BC019528.1 | - | 2 - 3 | 310 | 4208860 | 56 | |
BC031186.1 | - | 2 - 3 | 297 | 4989959 | 56 | |
BC036157.1 | - | 3 - 4 | 336 | 4982413 | 56 | |
BC036163.1 | - | 3 - 4 | 377 | 4984410 | 56 | |
BY096795.1 | - | 3 - 4 | 327 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |