Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145396.4 | NP_663371.2 | 17 - 18 | 1993 | 69 | |
GenBank mRNA | AK028806.1 | - | 17 - 18 | 1961 | 69 | |
AK081572.1 | - | 16 - 17 | 2051 | 69 | ||
AK081966.1 | - | 17 - 18 | 1994 | 69 | ||
AK083399.1 | - | 17 - 18 | 1976 | 69 | ||
AK137554.1 | - | 1962 | 69 | |||
AK149940.1 | - | 17 - 18 | 1972 | 69 | ||
AK163228.1 | - | 17 - 18 | 1963 | 69 | ||
AK165125.1 | - | 17 - 18 | 1963 | 69 | ||
AK167127.1 | - | 17 - 18 | 1967 | 69 | ||
AK218521.1 | - | 3 - 4 | 341 | 69 | ||
BC004624.1 | - | 7 - 8 | 913 | 69 | ||
BC019504.1 | - | 17 - 18 | 1949 | 4460615 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |