Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145528.3 | NP_663503.1 | 7 - 8 | 863 | 62 | |
GenBank mRNA | AK122339.1 | - | 3 - 4 | 582 | 62 | |
AK149672.1 | - | 7 - 8 | 822 | 62 | ||
AK154470.1 | - | 7 - 8 | 851 | 62 | ||
AK163962.1 | - | 7 - 8 | 816 | 62 | ||
AK171206.1 | - | 7 - 8 | 849 | 62 | ||
AK171440.1 | - | 7 - 8 | 840 | 62 | ||
BC016669.1 | - | 7 - 8 | 847 | 2648797 | 62 | |
BC023673.1 | - | 7 - 8 | 813 | 5343236 | 62 | |
BC023712.1 | - | 7 - 8 | 836 | 5346303 | 62 | |
BC033419.1 | - | 7 - 8 | 814 | 5359944 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |