Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020278.2 | NP_064674.1 | 3 - 4 | 461 | 132 | |
GenBank mRNA | AK031996.1 | - | 3 - 4 | 372 | 132 | |
AK045443.1 | - | 3 - 4 | 566 | 132 | ||
BC066090.1 | - | 3 - 4 | 461 | 6839091 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |