Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144919.2 | NP_659168.1 | 8 - 9 | 684 | 103 | |
GenBank mRNA | AK045994.1 | - | 8 - 9 | 696 | 103 | |
BC016208.1 | - | 8 - 9 | 684 | 4913322 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |