Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uroc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144940.2 | NP_659189.2 | 15 - 16 | 1539 | 114 | |
GenBank mRNA | AK149464.1 | - | 15 - 16 | 1642 | 114 | |
BC022133.1 | - | 15 - 16 | 1522 | 5098655 | 114 | |
BC033269.1 | - | 12 - 13 | 1154 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |