Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164612.1 | NP_001158084.1 | 9 - 10 | 1120 | 64 | |
NM_001164613.1 | NP_001158085.1 | 10 - 11 | 1173 | 64 | ||
NM_172613.4 | NP_766201.3 | 9 - 10 | 1120 | 64 | ||
GenBank mRNA | AK028776.1 | - | 10 - 11 | 1175 | 64 | |
AK029303.1 | - | 8 - 9 | 864 | 64 | ||
BC048410.1 | - | 9 - 10 | 1120 | 6486835 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |