Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172643.5 | NP_766231.2 | 9 - 10 | 2136 | 125 | |
GenBank mRNA | AK034756.1 | - | 8 - 9 | 1233 | 125 | |
AK036544.1 | - | 9 - 10 | 2135 | 125 | ||
AK037812.1 | - | 2 - 3 | 158 | 125 | ||
AK079719.1 | - | 5 - 6 | 494 | 125 | ||
AK163302.1 | - | 8 - 9 | 1409 | 125 | ||
BC046465.1 | - | 8 - 9 | 1305 | 125 | ||
BC150859.1 | - | 8 - 9 | 1475 | 9087770 | 125 | |
BC172012.1 | - | 9 - 10 | 1401 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |