Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172702.3 | NP_766290.2 | 1 - 2 | 132 | 72 | |
GenBank mRNA | AK075863.1 | - | 1 - 2 | 100 | 72 | |
BC031720.1 | - | 1 - 2 | 113 | 4209575 | 72 | |
BG077215.2 | - | 1 - 2 | 87 | 72 | ||
BU511121.1 | - | 1 - 2 | 68 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |