Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276287.1 | NP_001263216.1 | 3 - 4 | 457 | 57 | |
NM_181066.3 | NP_851411.1 | 3 - 4 | 359 | 57 | ||
NR_074085.1 | 3 - 4 | 359 | 57 | |||
GenBank mRNA | AK043212.1 | - | 3 - 4 | 322 | 57 | |
AK049321.1 | - | 3 - 4 | 348 | 57 | ||
AK084987.1 | - | 3 - 4 | 348 | 57 | ||
AK085091.1 | - | 3 - 4 | 348 | 57 | ||
AK154448.1 | - | 3 - 4 | 330 | 57 | ||
AK170269.1 | - | 3 - 4 | 351 | 57 | ||
BC068301.1 | - | 3 - 4 | 423 | 30061848 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |