Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040271.1 | - | 17 - 18 | 2570 | 75 | |
AK146866.1 | - | 2761 | 75 | |||
AK160810.1 | - | 17 - 18 | 2650 | 75 | ||
BC060678.1 | - | 16 - 17 | 2491 | 75 | ||
BC084587.1 | - | 17 - 18 | 2665 | 3602481 | 75 | |
EU380771.1 | - | 2787 | 75 | |||
EU380772.1 | - | 2845 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |