Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172854.2 | NP_766442.1 | 2 - 3 | 466 | 77 | |
GenBank mRNA | AK030069.1 | - | 287 | 77 | ||
AK035313.1 | - | 2 - 3 | 467 | 77 | ||
AK220202.1 | - | 1 - 2 | 277 | 77 | ||
BC137871.1 | - | 2 - 3 | 394 | 8860890 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |