Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173037.1 | NP_766625.1 | 14 - 15 | 2743 | 98 | |
GenBank mRNA | AK047206.1 | - | 9 - 10 | 1461 | 98 | |
AK077757.1 | - | 14 - 15 | 2728 | 98 | ||
AK085030.1 | - | 14 - 15 | 2743 | 98 | ||
BC066134.1 | - | 14 - 15 | 2674 | 98 | ||
BC066869.1 | - | 14 - 15 | 2673 | 30106406 | 98 | |
BC069838.1 | - | 14 - 15 | 2673 | 30106422 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |