Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202445.1 | NP_001189374.1 | 2 - 3 | 634 | 116 | |
NM_011162.5 | NP_035292.2 | 2 - 3 | 342 | 116 | ||
GenBank mRNA | AB221603.1 | - | 2 - 3 | 205 | 116 | |
AF003115.1 | - | 2 - 3 | 342 | 116 | ||
AF054611.1 | - | 2 - 3 | 342 | 116 | ||
AF109768.1 | - | 2 - 3 | 242 | 116 | ||
AF109769.1 | - | 2 - 3 | 305 | 116 | ||
AF109771.1 | - | 2 - 3 | 206 | 116 | ||
AF332075.1 | - | 2 - 3 | 205 | 116 | ||
AF332076.1 | - | 2 - 3 | 205 | 116 | ||
BC072578.1 | - | 2 - 3 | 526 | 6402485 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |