Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013867.2 | NP_038895.1 | 10 - 11 | 2485 | 69 | |
GenBank mRNA | AF179566.1 | - | 10 - 11 | 2433 | 69 | |
AK143894.1 | - | 10 - 11 | 2273 | 69 | ||
AK158241.1 | - | 10 - 11 | 2486 | 69 | ||
AK167578.1 | - | 10 - 11 | 2303 | 69 | ||
BC023930.1 | - | 10 - 11 | 2474 | 5324365 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |