Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174848.3 | NP_777273.2 | 2 - 3 | 1690 | 103 | |
GenBank mRNA | BC043118.1 | - | 2 - 3 | 1690 | 6408859 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |