Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175164.4 | NP_780373.3 | 15 - 16 | 1707 | 90 | |
GenBank mRNA | AK129176.1 | - | 15 - 16 | 1645 | 90 | |
BC141394.1 | - | 15 - 16 | 1576 | 9056007 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |