Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038993.3 | NP_001034082.1 | 7 - 8 | 1101 | 74 | |
NM_175201.5 | NP_780410.2 | 6 - 7 | 1131 | 74 | ||
GenBank mRNA | AK006888.1 | - | 6 - 7 | 1092 | 74 | |
AK147608.1 | - | 7 - 8 | 1158 | 74 | ||
AK163346.1 | - | 2 - 3 | 262 | 74 | ||
BC060730.1 | - | 6 - 7 | 1108 | 6408623 | 74 | |
BC062976.1 | - | 6 - 7 | 1045 | 6848803 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |