Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175319.4 | NP_780528.1 | 8 - 9 | 1225 | 66 | |
GenBank mRNA | AK049131.1 | - | 8 - 9 | 1226 | 66 | |
BC094908.1 | - | 8 - 9 | 1121 | 30021730 | 66 | |
BC132375.1 | - | 8 - 9 | 1212 | 40130652 | 66 | |
BC138307.1 | - | 8 - 9 | 1211 | 8861327 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |