Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 26 - 27 | 4785 | 66 | |
GenBank mRNA | AK030215.1 | - | 1 - 2 | 329 | 66 | |
AK087907.1 | - | 11 - 12 | 1701 | 66 | ||
AK220342.1 | - | 18 - 19 | 2347 | 66 | ||
BC068235.1 | - | 15 - 16 | 2852 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |