Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178395.3 | NP_848482.1 | 1 - 2 | 348 | 79 | |
GenBank mRNA | AK018284.1 | - | 320 | 79 | ||
AK030662.1 | - | 1 - 2 | 349 | 79 | ||
AK044652.1 | - | 1 - 2 | 342 | 79 | ||
AK046533.1 | - | 1 - 2 | 347 | 79 | ||
AK136296.1 | - | 1 - 2 | 347 | 79 | ||
AK195218.1 | - | 1 - 2 | 59 | 79 | ||
BC117761.1 | - | 2 - 3 | 110 | 79 | ||
BC127156.1 | - | 2 - 3 | 182 | 40090859 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |